(S)-(-)-1,2,3,4-Tetrahydro-1-naphthoic acid 98%

Supplier: Ambeed
Warning

Synonyms: (S)-1,2,3,4-Tetrahydro-1-naphthoic acid, (1S)-1,2,3,4-Tetrahydronaphthalene-1-carboxylic acid

A418479-1G A418479-25G A418479-5G A418479-10G
103827-354EA 13 USD
103827-354 103838-398 76841-998 76842-000 76842-002 76877-076
(S)-(-)-1,2,3,4-Tetrahydro-1-naphthoic acid 98%
(S)-(-)-1,2,3,4-Tetrahydro-1-naphthoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₂
MW: 176.22 g/mol
Boiling Pt: 342.7 °C (760 mm Hg)
Melting Pt: 54…56 °C
Flash Pt: 156.4 °C
Storage Temperature: Ambient
MDL Number: MFCD07368365
CAS Number: 85977-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR