(S)-(-)-2,2'-Diamino-1,1'-binaphthalene 98%

Supplier: Ambeed
Warning

Synonyms: (S)-1,1'-Binaphthyl-2,2'-diamine, (S)-(-)-BINAM, (S)-(-)-1,1'-Binaphthyl-2,2'-diamine, (S)-(-)-1,1'-Bi(2-naphthylamine), (S)-(-)-2,2'-Diamino-1,1'-binaphthyl

A480163-100G A480163-10G A480163-250MG
103828-116EA 9 USD
103828-116 103839-192 77004-576 77004-578 77004-580
(S)-(-)-2,2'-Diamino-1,1'-binaphthalene 98%
(S)-(-)-2,2'-Diamino-1,1'-binaphthalene

Precaución: for Research Use Only

Formula: C₂₀H₁₆N₂
MW: 284.362 g/mol
Boiling Pt: 480.8 °C (760 mm Hg)
Melting Pt: 242…244 °C
Flash Pt: 293.854 °C
Storage Temperature: Ambient
MDL Number: MFCD00145204
CAS Number: 18531-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR