(S)-1-Boc-piperidine-3-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-Boc-nipecotic acid, 1-Boc-L-nipecotic acid, (S)-1-(tert-Butoxycarbonyl)piperidine-3-carboxylic acid
A273936-100G
A273936-10G
A273936-25G
A273936-5G
A273936-1G
103813-166EA
5
USD
103813-166
103853-092
76802-038
76802-040
77092-906
77092-908
77305-152
(S)-1-Boc-piperidine-3-carboxylic acid 97%
(S)-1-Boc-piperidine-3-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₁H₁₉NO₄ MW: 229.28 g/mol Melting Pt: 165…169 °C Flash Pt: 167.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD02179172 CAS Number: 88495-54-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...