(S)-1-(tert-Butoxycarbonyl)azetidine-2-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: (S)-Azetidine-1,2-dicarboxylic acid 1-tert-butyl ester

A171324-25G A171324-5G A171324-100G A171324-10G A171324-250MG A171324-1G
103806-624EA 45 USD
103806-624 103844-898 77082-044 77082-046 77248-016 77248-018 77248-020 77301-122
(S)-1-(tert-Butoxycarbonyl)azetidine-2-carboxylic acid 97%
(S)-1-(tert-Butoxycarbonyl)azetidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₁₅NO₄
MW: 201.2200 g/mol
Boiling Pt: 321 °C (760 mm Hg)
Melting Pt: 105…110 °C
Flash Pt: 147.9 °C
Storage Temperature: Ambient
MDL Number: MFCD01321011
CAS Number: 51077-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR