(S)-1-(tert-Butoxycarbonyl)-4,4-difluoro-2-pyrrolidinecarboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: (2S)-1-[(tert-butoxy)carbonyl]-4,4-difluoropyrrolidine-2-carboxylic acid, (S)-1-(tert-Butoxycarbonyl)-4,4-difluoropyrrolidine-2-carboxylic acid, N-Boc-4,4-difluoro-L-proline, (S)-1-Boc-4,4-difluoro-2-pyrrolidinecarboxylic Acid, N-(tert-Butoxycarbonyl)-4,4-difluoro-L-proline

A141884-10G A141884-250MG A141884-1G A141884-25G A141884-5G
103803-072EA 19 USD
103803-072 77137-344 77137-346 77316-264 77316-266 77316-268
(S)-1-(tert-Butoxycarbonyl)-4,4-difluoro-2-pyrrolidinecarboxylic acid 97%
(S)-1-(tert-Butoxycarbonyl)-4,4-difluoro-2-pyrrolidinecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₅F₂NO₄
MW: 251.23 g/mol
Boiling Pt: 340.4 °C (760 mm Hg)
Melting Pt: 120…124 °C
Flash Pt: 159.7 °C
Storage Temperature: Ambient
MDL Number: MFCD03094917
CAS Number: 203866-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR