1-Boc-2-methyl-L-proline 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-1-(tert-Butoxycarbonyl)-2-methylpyrrolidine-2-carboxylic acid, Boc-α-Me-Pro-OH
A167224-10G
A167224-250MG
A167224-1G
A167224-5G
A167224-25G
A167224-100G
A167224-500G
103806-152EA
6
USD
103806-152
103844-414
76973-832
76973-834
77081-416
77081-418
77567-428
77671-364
77671-366
1-Boc-2-methyl-L-proline 97%
1-Boc-2-methyl-L-proline
Precaución: For research use only.
Formula:
C₁₁H₁₉NO₄ MW: 229.28 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD08706361 CAS Number: 103336-06-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...