(S)-1-(Pyridin-4-yl)ethanol 98%
Supplier: Ambeed
|
Warning
|
A134372-100MG
A134372-10G
A134372-250MG
A134372-25G
A134372-1G
103802-010EA
18
USD
103802-010
77137-028
77137-030
77137-032
77137-034
77314-756
(S)-1-(Pyridin-4-yl)ethanol 98%
(S)-1-(Pyridin-4-yl)ethanol
Precaución: for Research Use Only
Formula:
C₇H₉NO Melting Pt: 67…69 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00077866 CAS Number: 54656-96-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...