(S)-ɑ-Methyl-4-nitrobenzylamine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: (S)-Nitresolve, (S)-1-(4-Nitrophenyl)ethanamine hydrochloride, (S)-1-(4-Nitrophenyl)ethylamine Hydrochloride, (S)-(-)-α-Methyl-4-nitrobenzylamine hydrochloride

A129282-100G A129282-10G A129282-1G A129282-25G A129282-5G
103801-254EA 6 USD
103801-254 77190-008 77190-010 77313-634 77313-636 77313-638
(S)-ɑ-Methyl-4-nitrobenzylamine hydrochloride 98%
(S)-alpha-Methyl-4-nitrobenzylamine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂.HCl
MW: 202.64 g/mol
Melting Pt: 248…250 °C
Storage Temperature: Ambient
MDL Number: MFCD00066312
CAS Number: 132873-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR