(S)-4-Phenyl-3-[5-(4-fluorophenyl)-5-oxopentanoyl]-2-oxazolidinone 98%

Supplier: Ambeed
Warning

Synonyms: (S)-1-(4-Fluorophenyl)-5-(2-oxo-4-phenyloxazolidin-3-yl)pentane-1,5-dione

A186142-100G A186142-25G A186142-5G A186142-1G
103815-920EA 11 USD
103815-920 103846-494 76998-846 77084-146 77084-148 77302-026
(S)-4-Phenyl-3-[5-(4-fluorophenyl)-5-oxopentanoyl]-2-oxazolidinone 98%
(S)-4-Phenyl-3-[5-(4-fluorophenyl)-5-oxopentanoyl]-2-oxazolidinone

Precaución: for Research Use Only

Formula: C₂₀H₁₈FNO₄
MW: 355.37 g/mol
Boiling Pt: 568.4 °C (760 mm Hg)
Flash Pt: 297.6 °C
Storage Temperature: Ambient
MDL Number: MFCD08061377
CAS Number: 189028-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR