(S)-1-(4-Bromophenyl)propan-1-amine hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A259531-1G
A259531-100MG
A259531-250MG
A259531-50MG
103812-448EA
169
USD
103812-448
77091-898
77250-132
77250-134
77250-136
(S)-1-(4-Bromophenyl)propan-1-amine hydrochloride 97%
(S)-1-(4-Bromophenyl)propan-1-amine hydrochloride
Precaución: for Research Use Only
Formula:
C₁₄H₂₄O₃ Storage Temperature: Ambient |
MDL Number:
MFCD12910552 CAS Number: 1391577-97-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...