(S)-(-)-4-Bromo-α-methylbenzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: (S)-1-(4-Bromophenyl)ethanol

A218595-1G A218595-5G A218595-100MG A218595-10G
103818-722EA 26 USD
103818-722 103849-592 77088-118 77088-120 77139-810 77746-307
(S)-(-)-4-Bromo-α-methylbenzyl alcohol 98%
(S)-(-)-4-Bromo-α-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉BrO
Boiling Pt: 253.3 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD06201860
CAS Number: 100760-04-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR