(S)-(-)-4-Bromo-α-methylbenzylamine 98%

Supplier: Ambeed
Danger

Synonyms: (S)-1-(4-Bromophenyl)ethanamine, (S)-(-)-1-(4-Bromophenyl)ethylamine

A275477-10G A275477-1G A275477-5G A275477-25G
103813-260EA 5 USD
103813-260 103853-184 76772-218 77093-034 77093-036 77305-196
(S)-(-)-4-Bromo-α-methylbenzylamine 98%
(S)-(-)-4-Bromo-α-methylbenzylamine

Precaución: for Research Use Only

Formula: C₈H₁₀BrN
MW: 200.08 g/mol
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00066026
CAS Number: 27298-97-1
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR