(S)-1-[3,5-Bis(trifluoromethyl)phenyl]ethanol 96%

Supplier: Ambeed
Warning

Synonyms: (S)-1-[3,5-Bis(trifluoromethyl)phenyl]ethyl alcohol, (S)-(-)-3,5-Bis(trifluoromethyl)-alpha-methylbenzyl alcohol, (1S)-(-)-1-[3,5-Bis(trifluoromethyl)phenyl]ethan-1-ol, (S)-1-(3,5-Bis(trifluoromethyl)phenyl)ethanol, (S)-3,5-Bis(trifluoromethyl)-alpha-methylbenzyl alcohol

A131214-100G A131214-10G A131214-250MG A131214-5G A131214-1G A131214-25G
103799-336EA 7 USD
103799-336 103801-544 76797-878 76797-880 76797-882 77298-464 77314-020 77314-022
(S)-1-[3,5-Bis(trifluoromethyl)phenyl]ethanol 96%
(S)-1-[3,5-Bis(trifluoromethyl)phenyl]ethanol

Precaución: for Research Use Only

Formula: C₁₀H₈F₆O
MW: 258.16 g/mol
Boiling Pt: 223.5 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD03093011
CAS Number: 225920-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR