(S)-1-(Chloroacetyl)-2-pyrrolidinecarbonitrile 97%

Supplier: Ambeed
Warning

A166291-100G A166291-10G A166291-250MG A166291-1G
103806-046EA 6 USD
103806-046 103844-306 77138-314 77138-316 77138-318 77300-822
(S)-1-(Chloroacetyl)-2-pyrrolidinecarbonitrile 97%
(S)-1-(Chloroacetyl)-2-pyrrolidinecarbonitrile

Precaución: for Research Use Only

Formula: C₇H₉ClN₂O
Boiling Pt: 363.1 °C (760 mm Hg)
Flash Pt: 173.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08689902
CAS Number: 207557-35-5
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR