(S)-1-(2-Bromophenyl)ethanamine 97%

Supplier: Ambeed
Warning

A104713-100MG A104713-250MG
103796-764EA 59 USD
103796-764 103822-486 77135-652 77135-654
(S)-1-(2-Bromophenyl)ethanamine 97%
(S)-1-(2-Bromophenyl)ethanamine

Precaución: for Research Use Only

Formula: C₈H₁₀BrN
MW: 200.08 g/mol
Boiling Pt: 248.5 °C (760 mm Hg)
Melting Pt: 143…146 °C
Flash Pt: 104 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06761911
CAS Number: 140632-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR