(S)-1-((Benzyloxy)carbonyl)pyrrolidine-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A161242-10G A161242-25G A161242-5G
103805-470EA 29 USD
103805-470 103843-708 77191-076 77319-670 77319-672
(S)-1-((Benzyloxy)carbonyl)pyrrolidine-3-carboxylic acid 97%
(S)-1-((Benzyloxy)carbonyl)pyrrolidine-3-carboxylic acid

Precaución: Sealed in dry, at 2…8°C

Formula: C₁₃H₁₅NO₄
MW: 249.26 g/mol
Boiling Pt: 432.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09608053
CAS Number: 192214-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR