Fmoc-(S)-nipecotic acid 98%

Supplier: Ambeed
Warning

A218522-250MG A218522-1G A218522-5G
103818-714EA 33 USD
103818-714 103849-586 76999-840 77088-108 77088-110
Fmoc-(S)-nipecotic acid 98%
Fmoc-(S)-nipecotic acid

Precaución: for Research Use Only

Formula: C₂₁H₂₁NO₄
Boiling Pt: 561.6 °C (760 mm Hg)
Melting Pt: 160…161 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04112682
CAS Number: 193693-68-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR