(S)-(-)-1,1'-Bi-2-naphthol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-BINOL, (S)-(-)-1,1'-Binaphthyl-2,2'-Diol, (S)-[1,1'-Binaphthalene]-2,2'-diol
A436587-100G
A436587-10G
A436587-1G
A436587-500G
A436587-25G
103838-680EA
7
USD
103838-680
77291-568
77291-570
77291-572
77291-574
77307-294
(S)-(-)-1,1'-Bi-2-naphthol 98%
(S)-(-)-1,1'-Bi-2-naphthol
Precaución: Keep in a dark place, inert atmosphere, room temperature
Formula:
C₂₀H₁₄O₂ Melting Pt: 215…218 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004068 CAS Number: 18531-99-2 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...