(S)-1-Tetrahydrofuran-3-ylmethanamine 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
(S)-(Tetrahydrofuran-3-yl)methanamine
A471609-1G
A471609-100MG
A471609-250MG
103828-030EA
263
USD
103828-030
76843-012
76877-740
76877-742
(S)-1-Tetrahydrofuran-3-ylmethanamine 97%
(S)-1-Tetrahydrofuran-3-ylmethanamine
Precaución: for Research Use Only
Formula:
C₅H₁₁NO MW: 101.15 g/mol Boiling Pt: 156.0±13.0 °C (760 mm Hg) Flash Pt: 53.1 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD17015964 CAS Number: 1048962-84-0 UN: 2924 ADR: 3,8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...