(2S)-(+)-2-(Hydroxymethyl)tetrahydrofuran 96%

Supplier: Ambeed
Warning

Synonyms: (S)-(+)-Tetrahydrofurfuryl alcohol, (S)-(Tetrahydrofuran-2-yl)methanol, (2S)-(+)-(Tetrahydrofur-2-yl)methanol

A631787-100MG A631787-250MG
103840-276EA 115 USD
103840-276 77145-158 77145-160
(2S)-(+)-2-(Hydroxymethyl)tetrahydrofuran 96%
(2S)-(+)-2-(Hydroxymethyl)tetrahydrofuran

Precaución: for Research Use Only

Formula: C₅H₁₀O₂
MW: 102.132 g/mol
Boiling Pt: 176.8 °C (760 mm Hg)
Flash Pt: 83.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04972320
CAS Number: 57203-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR