(S)-(+)-Tetrahydrofurfurylamine 97%

Supplier: Ambeed
Danger

Synonyms: (S)-(+)-2-(Aminomethyl)tetrahydrofuran, (S)-Tetrahydrofurfurylamine, (S)-(Tetrahydrofuran-2-yl)methanamine

A101472-250MG A101472-25G A101472-1G A101472-5G
103795-990EA 43 USD
103795-990 103799-240 103821-678 77135-490 77135-492 77297-118 77297-120
(S)-(+)-Tetrahydrofurfurylamine 97%
(S)-(+)-Tetrahydrofurfurylamine

Precaución: for Research Use Only

Formula: C₅H₁₁NO
MW: 101.15 g/mol
Boiling Pt: 152…153 °C (760 mm Hg)
Flash Pt: 46 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00085303
CAS Number: 7175-81-7
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR