(S)-(+)-Pyrrolidine-3-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: (S)-(+)-Pyrrolidine-3-carboxylic Acid

A100954-250MG A100954-1G A100954-5G
103795-720EA 85 USD
103795-720 103821-410 77160-180 77282-620 77282-622
(S)-(+)-Pyrrolidine-3-carboxylic acid 95%
(S)-(+)-Pyrrolidine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₉NO₂
MW: 115.13 g/mol
Boiling Pt: 252.2 °C (760 mm Hg)
Melting Pt: 185…191 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06738463
CAS Number: 72580-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR