(S)-(+)-5-Oxotetrahydrofuran-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A692028-1G A692028-25G A692028-5G A692028-100G A692028-10G A692028-250MG
103840-588EA 18 USD
103840-588 76844-806 76844-808 76844-810 77005-942 77005-944 77005-946
(S)-(+)-5-Oxotetrahydrofuran-2-carboxylic acid 98%
(S)-(+)-5-Oxotetrahydrofuran-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₆O₄
Boiling Pt: 384.2 °C (760 mm Hg)
Melting Pt: 71…73 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064516
CAS Number: 21461-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR