(S)-(+)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol 98%

Supplier: Ambeed
Warning

A138506-100G A138506-10G A138506-500G A138506-1G A138506-25G A138506-5G
103802-620EA 5 USD
103802-620 76996-772 76996-774 76996-776 77315-652 77315-654 77315-656
(S)-(+)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol 98%
(S)-(+)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol

Precaución: for Research Use Only

Formula: C₆H₁₂O₃
Boiling Pt: 188.5 °C (760 mm Hg)
Flash Pt: 90 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063239
CAS Number: 22323-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR