(S)-(-)-α-Terpineol 97%

Supplier: Ambeed
Warning

Synonyms: (S)-2-(4-Methylcyclohex-3-en-1-yl)propan-2-ol

A405257-100G A405257-25G A405257-500G A405257-5G
103810-292EA 211 USD
103810-292 77252-154 77252-156 77252-158 77252-160
(S)-(-)-α-Terpineol 97%
(S)-(-)-α-Terpineol

Precaución: Sealed in dry, at 2…8°C

Formula: C₁₀H₁₈O
MW: 154.25
Boiling Pt: 217.5 °C (760 mm Hg)
Melting Pt: 31…35 °C
Flash Pt: 89.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00075926
CAS Number: 10482-56-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR