(S)-(-)-3-(N-Methylamino)-1-(2-thienyl)-1-propanol 97%

Supplier: Ambeed
Warning

Synonyms: (S)-2-[3-(Methylamino)-1-hydroxypropyl]thiophene, (S)-3-(Methylamino)-1-(2-thienyl)-1-propanol

A203951-1G A203951-5G
103817-512EA 5 USD
103817-512 103848-172 77086-318 77086-320
(S)-(-)-3-(N-Methylamino)-1-(2-thienyl)-1-propanol 97%
(S)-(-)-3-(N-Methylamino)-1-(2-thienyl)-1-propanol

Precaución: for Research Use Only

Formula: C₈H₁₃NOS
MW: 171.2600 g/mol
Boiling Pt: 294.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD07357308
CAS Number: 116539-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR