(S)-(-)-1-Methyl-2-pyrrolidinemethanol 97%

Supplier: Ambeed
Warning

Synonyms: (S)-N-Methylpyrrolidine-2-methanol, (S)-(-)-2-Hydroxymethyl-1-methylpyrrolidine, N-Methyl-L-prolinol

A245180-1G A245180-25G A245180-5G A245180-100G A245180-10G
103811-624EA 5 USD
103811-624 103851-460 77090-696 77090-698 77090-700 77288-840 77288-842
(S)-(-)-1-Methyl-2-pyrrolidinemethanol 97%
(S)-(-)-1-Methyl-2-pyrrolidinemethanol

Precaución: for Research Use Only

Formula: C₆H₁₃NO
MW: 115.176 g/mol
Flash Pt: 63 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011727
CAS Number: 34381-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR