(S)-(-)-1-(1-Naphthyl)ethylamine 98%

Supplier: Ambeed
Warning

Synonyms: (S)-(-)-alpha-(1-Naphthyl)ethylamine

A169876-100G A169876-10G A169876-500G A169876-1G A169876-25G A169876-5G
103806-424EA 7 USD
103806-424 103844-690 77028-988 77028-990 77028-992 77081-786 77081-788 77081-790
(S)-(-)-1-(1-Naphthyl)ethylamine 98%
(S)-(-)-1-(1-Naphthyl)ethylamine

Precaución: for Research Use Only

Formula: C₁₂H₁₃N
MW: 171.24 g/mol
Boiling Pt: 153 °C (760 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00064179
CAS Number: 10420-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR