(S)-(((1-(6-Amino-9H-purin-9-yl)propan-2-yl)oxy)methyl)phosphonic acid 95%
Supplier: Ambeed
|
Warning
|
A562467-1G
A562467-5G
A562467-10G
A562467-250MG
A562467-25G
103828-754EA
47
USD
103828-754
103839-888
76843-988
76843-990
76878-596
76878-598
76878-600
(S)-(((1-(6-Amino-9H-purin-9-yl)propan-2-yl)oxy)methyl)phosphonic acid 95%
(S)-(((1-(6-Amino-9H-purin-9-yl)propan-2-yl)oxy)methyl)phosphonic acid
Precaución: for Research Use Only
Formula:
C₉H₁₄N₅O₄P Boiling Pt: 616.1 °C (760 mm Hg) Flash Pt: 326.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD21604708 CAS Number: 147127-19-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...