(R,R)-(+)-Hydrobenzoin 97%

Supplier: Ambeed
Warning

Synonyms: (1R,2R)-1,2-Diphenylethane-1,2-diol, (R,R)-(+)-1,2-Diphenyl-1,2-ethanediol

A363247-5G A363247-10G A363247-250MG A363247-25G
103826-340EA 20 USD
103826-340 103837-360 76840-582 77142-340 77142-342 77142-344
(R,R)-(+)-Hydrobenzoin 97%
(R,R)-(+)-Hydrobenzoin

Precaución: for Research Use Only

Formula: C₁₄H₁₄O₂
MW: 214.26 g/mol
Boiling Pt: 373 °C (760 mm Hg)
Melting Pt: 146…149 °C
Flash Pt: 179.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00064254
CAS Number: 52340-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR