(R,E)-2-(4-(2,4-Dichlorophenyl)-1,3-dithiolan-2-ylidene)-2-(1H-imidazol-1-yl)acetonitrile ≥98%

Supplier: Ambeed
Warning

Synonyms: NND 502, Luliconazole, NND502, NND-502

A170757-250MG A170757-50MG A170757-10MG
103806-536EA 28 USD
103806-536 77600-382 77600-384 77600-380
(R,E)-2-(4-(2,4-Dichlorophenyl)-1,3-dithiolan-2-ylidene)-2-(1H-imidazol-1-yl)acetonitrile ≥98%
(R,E)-2-(4-(2,4-Dichlorophenyl)-1,3-dithiolan-2-ylidene)-2-(1H-imidazol-1-yl)acetonitrile

Precaución: Sealed in dry, store in freezer, under –20 °C.

Formula: C₁₄H₉Cl₂N₃S₂
MW: 354.27 g/mol
Boiling Pt: 499.1 °C (760 mm Hg)
Flash Pt: 255.6 °C
Storage Temperature: Freezer
MDL Number: MFCD00953915
CAS Number: 187164-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR