(R)-Tetrahydro-2H-pyran-3-amine hydrochloride 97%

Supplier: Ambeed
Warning

A120458-100MG A120458-250MG
103799-938EA 124 USD
103799-938 77026-384 77026-386
(R)-Tetrahydro-2H-pyran-3-amine hydrochloride 97%
(R)-Tetrahydro-2H-pyran-3-amine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₂ClNO
MW: 137.6 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09056863
CAS Number: 1315500-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR