D-Proline-tert-butyl ester hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: H-D-Pro-OtBu.HCl, H-D-Pro-OtBu·HCl, (R)-tert-Butyl pyrrolidine-2-carboxylate hydrochloride

A151222-100MG A151222-10G A151222-250MG A151222-1G A151222-25G A151222-5G A151222-100G
103804-236EA 17 USD
103804-236 103842-384 77010-790 77010-792 77010-794 77299-856 77299-858 77299-860 77683-275
D-Proline-tert-butyl ester hydrochloride 98%
D-Proline-tert-butyl ester hydrochloride

Precaución: For research use only.

Formula: C₉H₁₇NO₂
MW: 207.70 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00153460
CAS Number: 184719-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR