(3R)-(+)-3-(tert-Butoxycarbonylamino)pyrrolidine 98%

Supplier: Ambeed
Warning

Synonyms: (3R)-(+)-3-(Boc-amino)pyrrolidine, (R)-tert-Butyl pyrrolidin-3-ylcarbamate

A271593-100G A271593-10G A271593-1G A271593-25G A271593-5G
103813-050EA 6 USD
103813-050 103852-964 76692-846 76692-848 77092-740 77092-742 77092-744
(3R)-(+)-3-(tert-Butoxycarbonylamino)pyrrolidine 98%
(3R)-(+)-3-(tert-Butoxycarbonylamino)pyrrolidine

Precaución: for Research Use Only

Formula: C₉H₁₈N₂O₂
MW: 186.26 g/mol
Flash Pt: 127 °C
Storage Temperature: Ambient
MDL Number: MFCD00143191
CAS Number: 122536-77-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR