(R)-tert-Butyl methyl(pyrrolidin-3-yl)carbamate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(3R)-3-(Methylamino)pyrrolidine, 3-BOC protected
A126630-250MG
A126630-1G
A126630-25G
103800-844EA
25
USD
103800-844
76810-948
77313-072
77313-074
(R)-tert-Butyl methyl(pyrrolidin-3-yl)carbamate 97%
(R)-tert-Butyl methyl(pyrrolidin-3-yl)carbamate
Precaución: for Research Use Only
Formula:
C₁₀H₂₀N₂O₂ Boiling Pt: 268 °C (760 mm Hg) Flash Pt: 116 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09263387 CAS Number: 392338-15-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...