(R)-tert-Butyl methyl(piperidin-3-yl)carbamate 95%

Supplier: Ambeed
Warning

A278872-1G A278872-10G A278872-250MG
103813-430EA 26 USD
103813-430 103853-374 77093-298 77099-138 77099-140
(R)-tert-Butyl methyl(piperidin-3-yl)carbamate 95%
(R)-tert-Butyl methyl(piperidin-3-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
Boiling Pt: 287.8 °C (760 mm Hg)
Flash Pt: 128 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09878596
CAS Number: 309962-67-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR