(R)-tert-Butyl azepan-3-ylcarbamate 98%

Supplier: Ambeed
Warning

A140619-100MG A140619-250MG
103802-888EA 195 USD
103802-888 76647-020 76647-022
(R)-tert-Butyl azepan-3-ylcarbamate 98%
(R)-tert-Butyl azepan-3-ylcarbamate

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
MW: 214.3 g/mol
Boiling Pt: 322.7±31.0 °C (760 mm Hg)
Flash Pt: 149 °C
Storage Temperature: Refrigerator
MDL Number: MFCD19686115
CAS Number: 1354351-56-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR