(R)-1-Boc-3-Hydroxypiperidine 97%

Supplier: Ambeed
Warning

Synonyms: (R)-1-BOC-3-hydroxypiperidine, (R)-tert-Butyl 3-hydroxypiperidine-1-carboxylate, tert-butyl (3R)-3-hydroxypiperidine-1-carboxylate

A185071-25G A185071-5G A185071-1G A185071-10G A185071-100G
103815-816EA 5 USD
103815-816 103846-382 77084-016 77084-018 77301-944 77567-696 77625-878
(R)-1-Boc-3-Hydroxypiperidine 97%
(R)-1-Boc-3-hydroxypiperidine

Precaución: For research use only.

Formula: C₁₀H₁₉NO₃
MW: 201.27 g/mol
Melting Pt: 43…50 °C
Flash Pt: 104.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04115306
CAS Number: 143900-43-0
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR