(R)-1-Boc-3-Ethylpiperazine 98%

Supplier: Ambeed
Warning

Synonyms: (R)-tert-Butyl 3-ethylpiperazine-1-carboxylate, (3R)-3-Ethylpiperazine, N1-BOC protected

A358926-1G A358926-5G A358926-100MG A358926-250MG
103826-214EA 43 USD
103826-214 103837-222 76840-382 76840-384 77106-888 77106-890
(R)-1-Boc-3-Ethylpiperazine 98%
(R)-1-Boc-3-ethylpiperazine

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
MW: 214.3100 g/mol
Boiling Pt: 286.7 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD07772111
CAS Number: 438050-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR