(R)-1-Boc-3-Cyanopyrrolidine 95%

Supplier: Ambeed
Warning

Synonyms: (R)-tert-Butyl 3-cyanopyrrolidine-1-carboxylate

A164572-10G A164572-250MG A164572-1G
103805-884EA 51 USD
103805-884 103844-138 77151-034 77151-036 77300-726
(R)-1-Boc-3-Cyanopyrrolidine 95%
(R)-1-Boc-3-Cyanopyrrolidine

Precaución: for Research Use Only

Formula: C₁₀H₁₆N₂O₂
MW: 196.2500 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05863904
CAS Number: 132945-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR