(R)-tert-Butyl 3-(tosyloxy)pyrrolidine-1-carboxylate 95%

Supplier: Ambeed
Warning

A145459-100MG A145459-250MG A145459-1G A145459-5G
103803-538EA 20 USD
103803-538 103841-624 77162-552 77162-554 77316-924 77316-926
(R)-tert-Butyl 3-(tosyloxy)pyrrolidine-1-carboxylate 95%
(R)-tert-Butyl 3-(tosyloxy)pyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₆H₂₃NO₅S
MW: 341.42 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16621198
CAS Number: 139986-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR