(R)-tert-Butyl-3-(hydroxymethyl)pyrrolidine-1-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: [R]-1-BOC-3-Pyrrolidinemethanol, (R)-tert-Butyl 3-(hydroxymethyl)pyrrolidine-1-carboxylate

A257926-10G A257926-250MG A257926-1G A257926-25G A257926-5G A257926-100G
103812-348EA 11 USD
103812-348 103852-222 77014-114 77014-116 77091-746 77091-748 77304-724 77550-628
(R)-tert-Butyl-3-(hydroxymethyl)pyrrolidine-1-carboxylate 97%
(R)-tert-Butyl-3-(hydroxymethyl)pyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₃
MW: 201.27 g/mol
Boiling Pt: 289.5 °C (760 mm Hg)
Flash Pt: 128.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03094725
CAS Number: 138108-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR