(R)-tert-Butyl-3-(hydroxymethyl)piperidine-1-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: (R)-1-BOC-3-(hydroxymethyl)piperidine, (R)-tert-Butyl 3-(hydroxymethyl)piperidine-1-carboxylate, 3-(R)-N-BOC-PIPERIDINE METHANOL, 1-Piperidinecarboxylicacid, 3-(hydroxymethyl)-, 1,1-dimethylethyl ester, (3R)-

A131793-100MG A131793-10G A131793-250MG A131793-1G
103801-632EA 12 USD
103801-632 76797-920 76797-922 76797-924 77314-158
(R)-tert-Butyl-3-(hydroxymethyl)piperidine-1-carboxylate 97%
(R)-tert-Butyl-3-(hydroxymethyl)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₁NO₃
MW: 215.293 g/mol
Boiling Pt: 308 °C (760 mm Hg)
Melting Pt: 77…81 °C
Flash Pt: 140.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02683202
CAS Number: 140695-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR