(R)-1-Boc-3-(Hydroxymethyl)piperazine 95%
Supplier: Ambeed
|
Warning
|
A112509-10G
A112509-25G
A112509-1G
A112509-5G
A112509-100G
103798-024EA
6
USD
103798-024
103823-860
77094-052
77094-054
77310-668
77310-670
77610-892
(R)-1-Boc-3-(Hydroxymethyl)piperazine 95%
(R)-1-Boc-3-(hydroxymethyl)piperazine
Precaución: for Research Use Only
Formula:
C₁₀H₂₀N₂O₃ Boiling Pt: 322.9 °C (760 mm Hg) Melting Pt: 68…71 °C Flash Pt: 149.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD05863888 CAS Number: 278788-66-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...