(R)-(-)-1-Boc-3-(aminomethyl)pyrrolidine 97%

Supplier: Ambeed
Warning

Synonyms: (3R)-3-(Aminomethyl)pyrrolidine, N1-BOC protected, (R)-tert-Butyl 3-(aminomethyl)pyrrolidine-1-carboxylate

A126014-100MG A126014-250MG A126014-1G
103800-750EA 63 USD
103800-750 76688-402 76688-404 77312-916
(R)-(-)-1-Boc-3-(aminomethyl)pyrrolidine 97%
(R)-(-)-1-Boc-3-(aminomethyl)pyrrolidine

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₂
MW: 200.282 g/mol
Boiling Pt: 280.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD03844734
CAS Number: 199174-29-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR