(R)-1-Boc-2-Methylpiperazine 97%

Supplier: Ambeed
Warning

Synonyms: (2R)-2-Methylpiperazine, N1-BOC protected, (R)-tert-Butyl 2-methylpiperazine-1-carboxylate

A202259-100G A202259-10G A202259-1G A202259-5G A202259-25G
103817-368EA 14 USD
103817-368 103848-004 76691-246 76691-248 77086-106 77086-108 77302-800
(R)-1-Boc-2-Methylpiperazine 97%
(R)-1-Boc-2-methylpiperazine

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD01862120
CAS Number: 170033-47-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR