(R)-tert-Butyl 2-methyl-4-oxopiperidine-1-carboxylate 98%

Supplier: Ambeed
Warning

A247089-10G A247089-250MG A247089-1G A247089-5G
103811-748EA 15 USD
103811-748 103851-580 77031-382 77031-384 77090-888 77090-890
(R)-tert-Butyl 2-methyl-4-oxopiperidine-1-carboxylate 98%
(R)-tert-Butyl 2-methyl-4-oxopiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₃
MW: 213.27 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09832896
CAS Number: 790667-43-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR