tert-Butyl-(R)-(+)-2-hydroxybutyrate 95%

Supplier: Ambeed
Warning

Synonyms: (R)-tert-Butyl 2-hydroxybutanoate

A135965-100MG A135965-10G A135965-250MG A135965-1G A135965-5G
103802-252EA 197 USD
103802-252 76782-658 76782-660 76782-662 77315-134 77315-136
tert-Butyl-(R)-(+)-2-hydroxybutyrate 95%
tert-Butyl-(R)-(+)-2-hydroxybutyrate

Precaución: for Research Use Only

Formula: C₈H₁₆O₃
MW: 160.211 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00210055
CAS Number: 206996-51-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR