(R)-tert-Butyl-2-(hydroxymethyl)morpholine-4-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: (R)-N-Boc-2-hydroxymethylmorpholine, (R)-tert-Butyl 2-(hydroxymethyl)morpholine-4-carboxylate

A334933-25G A334933-5G A334933-100G A334933-10G A334933-250MG A334933-500G A334933-1G
103825-674EA 9 USD
103825-674 103836-644 76839-650 76839-652 77141-990 77141-992 77141-994 77141-996 77306-478
(R)-tert-Butyl-2-(hydroxymethyl)morpholine-4-carboxylate 98%
(R)-tert-Butyl-2-(hydroxymethyl)morpholine-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₄
MW: 217.27 g/mol
Boiling Pt: 321 °C (760 mm Hg)
Flash Pt: 148 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09260605
CAS Number: 135065-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR