tert-Butyl N-[(3R)-piperidin-3-ylmethyl]carbamate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-tert-Butyl (piperidin-3-ylmethyl)carbamate
A170352-10G
A170352-1G
A170352-5G
103806-476EA
12
USD
103806-476
103844-752
76973-968
77301-044
77301-046
tert-Butyl N-[(3R)-piperidin-3-ylmethyl]carbamate 97%
tert-Butyl N-[(3R)-piperidin-3-ylmethyl]carbamate
Precaución: for Research Use Only
Formula:
C₁₁H₂₂N₂O₂ MW: 214.305 g/mol Boiling Pt: 321.8±15.0 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD03093384 CAS Number: 879275-33-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...